Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HESX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015446088.1 | XP_015301574.1 | 2 - 3 | 855 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |