Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567584.2 | XP_005567641.1 | 1 - 2 | 1427 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |