Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102120532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579495.2 | XP_005579552.1 | 9 - 10 | 1518 | 108 | |
XM_015435645.1 | XP_015291131.1 | 8 - 9 | 2049 | 108 | ||
XM_015435646.1 | XP_015291132.1 | 8 - 9 | 2047 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |