Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557353.2 | XP_005557410.1 | 3 - 4 | 433 | 136 | |
XM_005557354.2 | XP_005557411.1 | 3 - 4 | 395 | 136 | ||
XM_005557355.2 | XP_005557412.1 | 3 - 4 | 392 | 136 | ||
XM_005557356.2 | XP_005557413.1 | 3 - 4 | 378 | 136 | ||
XM_005557357.2 | XP_005557414.1 | 3 - 4 | 406 | 136 | ||
XM_005557358.2 | XP_005557415.1 | 3 - 4 | 350 | 136 | ||
XM_015451684.1 | XP_015307170.1 | 3 - 4 | 407 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |