Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558903.2 | XP_005558960.1 | 4 - 5 | 715 | 51 | |
XM_015452506.1 | XP_015307992.1 | 5 - 6 | 775 | 51 | ||
XM_015452507.1 | XP_015307993.1 | 4 - 5 | 725 | 51 | ||
XM_015452508.1 | XP_015307994.1 | 5 - 6 | 765 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |