Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110204.1 | NP_001103674.1 | 9 - 10 | 1461 | 68 | |
NM_001110205.1 | NP_001103675.1 | 7 - 8 | 939 | 68 | ||
NM_007394.3 | NP_031420.2 | 8 - 9 | 1216 | 68 | ||
GenBank mRNA | AF332087.1 | - | 6 - 7 | 849 | 68 | |
AF332088.1 | - | 6 - 7 | 849 | 68 | ||
AK052246.1 | - | 1051 | 68 | |||
AK053428.1 | - | 9 - 10 | 1460 | 68 | ||
AK150014.1 | - | 8 - 9 | 1216 | 68 | ||
AK150075.1 | - | 8 - 9 | 1216 | 68 | ||
BC058718.1 | - | 6 - 7 | 937 | 6418954 | 68 | |
L15436.1 | - | 7 - 8 | 1289 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |