Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007938.2 | NP_031964.2 | 3 - 4 | 1400 | 99 | |
GenBank mRNA | BC141090.1 | - | 3 - 4 | 916 | 9055702 | 99 |
BC145362.1 | - | 4 - 5 | 1036 | 99 | ||
U58332.1 | - | 3 - 4 | 1116 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |