Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001294138.1 | NP_001281067.1 | 3 - 4 | 578 | 127 | |
NM_001294140.1 | NP_001281069.1 | 3 - 4 | 578 | 127 | ||
NM_008194.3 | NP_032220.1 | 3 - 4 | 578 | 127 | ||
NM_212444.2 | NP_997609.1 | 3 - 4 | 578 | 127 | ||
GenBank mRNA | AK008186.1 | - | 3 - 4 | 382 | 127 | |
AK044308.1 | - | 3 - 4 | 416 | 127 | ||
AK088373.1 | - | 3 - 4 | 397 | 127 | ||
AK169424.1 | - | 3 - 4 | 384 | 127 | ||
AK169525.1 | - | 3 - 4 | 579 | 127 | ||
BC003767.1 | - | 3 - 4 | 381 | 3593896 | 127 | |
U48403.1 | - | 3 - 4 | 355 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |