Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008359.2 | NP_032385.1 | 11 - 12 | 1189 | 91 | |
GenBank mRNA | AK036587.1 | - | 11 - 12 | 1140 | 91 | |
AK080226.2 | - | 9 - 10 | 2022 | 91 | ||
U31993.1 | - | 11 - 12 | 1177 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |