Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243072.1 | NP_001230001.1 | 5 - 6 | 835 | 83 | |
NM_001243073.1 | NP_001230002.1 | 5 - 6 | 675 | 83 | ||
NM_009152.4 | NP_033178.2 | 4 - 5 | 1103 | 83 | ||
GenBank mRNA | AK140817.1 | - | 5 - 6 | 676 | 83 | |
AK164242.1 | - | 7 - 8 | 878 | 83 | ||
BC057588.1 | - | 4 - 5 | 750 | 6416990 | 83 | |
BC090844.1 | - | 5 - 6 | 835 | 30532393 | 83 | |
D85028.2 | - | 4 - 5 | 1105 | 83 | ||
L40484.1 | - | 1 - 2 | 136 | 83 | ||
L41541.1 | - | 4 - 5 | 454 | 83 | ||
X85993.1 | - | 4 - 5 | 564 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |