Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164480.1 | NP_001157952.1 | 11 - 12 | 2778 | 61 | |
NM_001164481.1 | NP_001157953.1 | 12 - 13 | 3032 | 61 | ||
NM_001164482.1 | NP_001157954.1 | 10 - 11 | 2717 | 61 | ||
NM_001164568.1 | NP_001158040.1 | 10 - 11 | 2747 | 61 | ||
NM_011379.4 | NP_035509.4 | 11 - 12 | 3001 | 61 | ||
GenBank mRNA | AK028151.1 | - | 10 - 11 | 1988 | 61 | |
AK137578.1 | - | 11 - 12 | 2724 | 61 | ||
AK149669.1 | - | 11 - 12 | 2724 | 61 | ||
AK156280.1 | - | 12 - 13 | 2986 | 61 | ||
AK169945.1 | - | 10 - 11 | 2663 | 61 | ||
AK220472.1 | - | 11 - 12 | 2914 | 61 | ||
BC043679.1 | - | 3 - 4 | 1138 | 61 | ||
BC054824.1 | - | 11 - 12 | 2722 | 5324326 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |