Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011930.3 | NP_036060.1 | 16 - 17 | 1521 | 60 | |
GenBank mRNA | AK030444.1 | - | 16 - 17 | 1522 | 60 | |
AK032674.1 | - | 3 - 4 | 251 | 60 | ||
AK128981.1 | - | 1489 | 60 | |||
AK150395.1 | - | 16 - 17 | 1432 | 60 | ||
BC050907.1 | - | 16 - 17 | 1493 | 5698121 | 60 | |
BC053049.1 | - | 16 - 17 | 1493 | 5698121 | 60 | |
BC054799.1 | - | 16 - 17 | 1501 | 6849205 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |