Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013477.3 | NP_038505.2 | 5 - 6 | 768 | 74 | |
GenBank mRNA | AB088408.1 | - | 5 - 6 | 657 | 74 | |
AK083372.1 | - | 5 - 6 | 769 | 74 | ||
AK133921.1 | - | 5 - 6 | 776 | 74 | ||
AK171515.1 | - | 5 - 6 | 738 | 74 | ||
BC011075.1 | - | 5 - 6 | 659 | 3662404 | 74 | |
U13840.1 | - | 5 - 6 | 657 | 74 | ||
U21549.1 | - | 5 - 6 | 640 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |