Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163489.1 | NP_001156961.1 | 6 - 7 | 630 | 60 | |
NM_001163490.1 | NP_001156962.1 | 6 - 7 | 709 | 60 | ||
NM_001163491.1 | NP_001156963.1 | 6 - 7 | 668 | 60 | ||
NM_013658.3 | NP_038686.3 | 6 - 7 | 808 | 60 | ||
GenBank mRNA | AK035918.1 | - | 5 - 6 | 670 | 60 | |
AK077976.1 | - | 6 - 7 | 788 | 60 | ||
AK152728.1 | - | 6 - 7 | 800 | 60 | ||
AK153164.1 | - | 6 - 7 | 800 | 60 | ||
AK154392.1 | - | 6 - 7 | 769 | 60 | ||
BC025800.1 | - | 6 - 7 | 726 | 5102015 | 60 | |
X85991.1 | - | 6 - 7 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |