Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900003853.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003853.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003853.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003853.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003853.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003851.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003851.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003851.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003851.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003851.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003857.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003857.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003857.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003857.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003857.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003860.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003860.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003860.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003860.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003860.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003852.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003852.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003852.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003852.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003852.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003859.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003859.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003859.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003859.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003859.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003861.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003861.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003861.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003861.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003861.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003850.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003850.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003850.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003850.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003850.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003854.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003854.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003854.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003854.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003854.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003855.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003855.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003855.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003855.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003855.mm.1 |
TC1900000415.mm.1 |
MTA TC |