Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199015.1 | NP_001185944.1 | 8 - 9 | 958 | 100 | |
NM_001199016.1 | NP_001185945.1 | 7 - 8 | 846 | 100 | ||
NM_021291.3 | NP_067266.1 | 7 - 8 | 1073 | 100 | ||
GenBank mRNA | AF192310.1 | - | 8 - 9 | 958 | 100 | |
AJ249198.1 | - | 7 - 8 | 1073 | 100 | ||
AK142541.1 | - | 7 - 8 | 847 | 100 | ||
AK164714.1 | - | 6 - 7 | 1263 | 100 | ||
BC010746.1 | - | 8 - 9 | 936 | 4236578 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |