Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021485.2 | NP_067460.1 | 1 - 2 | 74 | 63 | |
GenBank mRNA | AJ007938.1 | - | 1 - 2 | 74 | 63 | |
AK014412.1 | - | 1 - 2 | 153 | 63 | ||
AK090093.1 | - | 129 | 63 | |||
AK156361.1 | - | 1 - 2 | 127 | 63 | ||
BC051631.1 | - | 1 - 2 | 129 | 63 | ||
BC066842.1 | - | 1 - 2 | 145 | 30468251 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |