Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ezr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009510.2 | NP_033536.2 | 7 - 8 | 935 | 87 | |
GenBank mRNA | AK002766.1 | - | 7 - 8 | 909 | 87 | |
AK004588.2 | - | 7 - 8 | 910 | 87 | ||
AK035271.1 | - | 3 - 4 | 303 | 87 | ||
AK135194.1 | - | 7 - 8 | 911 | 87 | ||
AK135353.1 | - | 7 - 8 | 911 | 87 | ||
AK145707.1 | - | 7 - 8 | 936 | 87 | ||
AK159717.1 | - | 7 - 8 | 913 | 87 | ||
AK159824.1 | - | 7 - 8 | 911 | 87 | ||
AK159937.1 | - | 7 - 8 | 912 | 87 | ||
AK166191.1 | - | 7 - 8 | 911 | 87 | ||
AK168840.1 | - | 7 - 8 | 911 | 87 | ||
AK170610.1 | - | 7 - 8 | 911 | 87 | ||
AK172336.1 | - | 7 - 8 | 918 | 87 | ||
BC048181.1 | - | 7 - 8 | 898 | 6826190 | 87 | |
BC098502.1 | - | 7 - 8 | 887 | 30047137 | 87 | |
X60671.1 | - | 7 - 8 | 893 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |