Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286037.1 | NP_001272966.1 | 1 - 2 | 128 | 70 | |
NM_010876.4 | NP_035006.3 | 1 - 2 | 128 | 70 | ||
NR_104383.1 | 1 - 2 | 128 | 70 | |||
GenBank mRNA | AB002663.1 | - | 1 - 2 | 107 | 70 | |
AK149619.1 | - | 1 - 2 | 129 | 70 | ||
AK149668.1 | - | 1 - 2 | 95 | 70 | ||
AK149732.1 | - | 1 - 2 | 129 | 70 | ||
AK150945.1 | - | 1 - 2 | 94 | 70 | ||
AK151316.1 | - | 1 - 2 | 95 | 70 | ||
AK151593.1 | - | 1 - 2 | 93 | 70 | ||
AK152386.1 | - | 1 - 2 | 91 | 70 | ||
AK152992.1 | - | 1 - 2 | 95 | 70 | ||
AK162308.1 | - | 1 - 2 | 95 | 70 | ||
AK162623.1 | - | 1 - 2 | 98 | 70 | ||
AK170462.1 | - | 1 - 2 | 95 | 70 | ||
AK171559.1 | - | 1 - 2 | 94 | 70 | ||
BC055836.1 | - | 1 - 2 | 82 | 3983481 | 70 | |
L11455.1 | - | 1 - 2 | 65 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |