Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011703.4 | NP_035833.2 | 4 - 5 | 535 | 73 | |
GenBank mRNA | AF266282.1 | - | 4 - 5 | 417 | 73 | |
AK052465.1 | - | 4 - 5 | 536 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Probe spans exons |