Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018768.2 | NP_061238.1 | 1 - 2 | 36 | 66 | |
GenBank mRNA | AB040054.1 | - | 1 - 2 | 33 | 66 | |
AF093064.1 | - | 1 - 2 | 45 | 66 | ||
AK003297.1 | - | 1 - 2 | 62 | 66 | ||
AK011376.1 | - | 1 - 2 | 50 | 66 | ||
AK016271.1 | - | 1 - 2 | 62 | 66 | ||
AK035105.1 | - | 1 - 2 | 40 | 66 | ||
BC048167.1 | - | 1 - 2 | 36 | 6821351 | 66 | |
BC048479.1 | - | 1 - 2 | 61 | 6774827 | 66 | |
BC061118.1 | - | 1 - 2 | 61 | 30295878 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |