Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022653.4 | NP_073144.3 | 11 - 12 | 1957 | 77 | |
GenBank mRNA | AF314187.1 | - | 11 - 12 | 1915 | 77 | |
AK028609.1 | - | 11 - 12 | 1891 | 77 | ||
BC031175.1 | - | 3 - 4 | 468 | 77 | ||
BC031722.1 | - | 11 - 12 | 1923 | 4019169 | 77 | |
BC039651.1 | - | 3 - 4 | 468 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |