Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Robo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF536772.1 | - | 10 - 11 | 1592 | 74 | |
AK004723.2 | - | 10 - 11 | 1589 | 74 | ||
AK030973.1 | - | 379 | 74 | |||
AK087355.1 | - | 9 - 10 | 1742 | 74 | ||
AK172104.1 | - | 9 - 10 | 1830 | 74 | ||
BC020129.1 | - | 2 - 3 | 133 | 74 | ||
BC071193.1 | - | 9 - 10 | 1282 | 5060473 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |