Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172098.1 | NP_001165569.1 | 10 - 11 | 1557 | 129 | |
NM_052973.2 | NP_443205.1 | 12 - 13 | 1809 | 129 | ||
GenBank mRNA | AF307777.1 | - | 12 - 13 | 1793 | 129 | |
AK140447.1 | - | 10 - 11 | 1545 | 129 | ||
AK165908.1 | - | 10 - 11 | 1241 | 129 | ||
BC138055.1 | - | 10 - 11 | 1508 | 8861074 | 129 | |
BC145127.1 | - | 10 - 11 | 1508 | 9053675 | 129 | |
EF685152.1 | - | 13 - 14 | 1710 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |