Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007799.3 | NP_031825.2 | 4 - 5 | 714 | 70 | |
GenBank mRNA | AK143581.1 | - | 3 - 4 | 416 | 70 | |
AK145875.1 | - | 4 - 5 | 571 | 70 | ||
AK157907.1 | - | 3 - 4 | 436 | 70 | ||
AK165242.1 | - | 3 - 4 | 436 | 70 | ||
AK165271.1 | - | 3 - 4 | 382 | 70 | ||
BC005432.1 | - | 4 - 5 | 714 | 3601519 | 70 | |
CT010252.1 | - | 3 - 4 | 350 | 70 | ||
X97399.1 | - | 3 - 4 | 401 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |