Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163024.1 | NP_001156496.1 | 18 - 19 | 2805 | 121 | |
NM_001163025.1 | NP_001156497.1 | 18 - 19 | 2808 | 121 | ||
NM_153395.2 | NP_700444.2 | 18 - 19 | 2805 | 121 | ||
GenBank mRNA | AB014059.1 | - | 18 - 19 | 2327 | 121 | |
AK122425.1 | - | 18 - 19 | 2805 | 121 | ||
AK163740.1 | - | 6 - 7 | 2649 | 121 | ||
BC048406.1 | - | 5 - 6 | 581 | 121 | ||
BC055354.1 | - | 1 - 2 | 51 | 121 | ||
BC150943.1 | - | 18 - 19 | 2329 | 9087854 | 121 | |
CX122556.1 | - | 5 - 6 | 419 | 121 | ||
CX224591.1 | - | 3 - 4 | 199 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |