Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153589.2 | NP_705817.2 | 8 - 9 | 1255 | 102 | |
GenBank mRNA | AK044763.1 | - | 8 - 9 | 1256 | 102 | |
BC033409.1 | - | 7 - 8 | 850 | 5357763 | 102 | |
BC060368.1 | - | 7 - 8 | 859 | 102 | ||
FJ384096.1 | - | 8 - 9 | 1256 | 102 | ||
KC164759.2 | - | 8 - 9 | 1255 | 102 | ||
KC164760.2 | - | 7 - 8 | 1156 | 102 | ||
KC164761.2 | - | 7 - 8 | 948 | 102 | ||
KC164762.2 | - | 6 - 7 | 849 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |