Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252447.1 | NP_001239376.1 | 10 - 11 | 103 | ||
NM_027260.3 | NP_081536.2 | 10 - 11 | 103 | |||
GenBank mRNA | AF513620.1 | - | 9 - 10 | 103 | ||
AK012664.1 | - | 10 - 11 | 103 | |||
AK154485.1 | - | 10 - 11 | 103 | |||
BC013520.1 | - | 9 - 10 | 4236402 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |