Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133918.2 | NP_598679.1 | 4 - 5 | 2533 | 100 | |
GenBank mRNA | AK029337.1 | - | 4 - 5 | 2504 | 100 | |
AK052368.1 | - | 4 - 5 | 2613 | 100 | ||
AK088995.1 | - | 4 - 5 | 2598 | 100 | ||
AK140388.1 | - | 2 - 3 | 1949 | 100 | ||
AK155486.1 | - | 4 - 5 | 2835 | 100 | ||
BC005481.1 | - | 4 - 5 | 2533 | 3485057 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |