Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010423.2 | NP_034553.2 | 1 - 2 | 258 | 93 | |
GenBank mRNA | AB041590.1 | - | 1 - 2 | 151 | 93 | |
AF151521.1 | - | 1 - 2 | 206 | 93 | ||
AF172286.1 | - | 1 - 2 | 239 | 93 | ||
AF176423.1 | - | 1 - 2 | 220 | 93 | ||
AF232241.1 | - | 1 - 2 | 238 | 93 | ||
AJ243895.1 | - | 1 - 2 | 206 | 93 | ||
AK140212.1 | - | 1 - 2 | 259 | 93 | ||
BC086635.1 | - | 1 - 2 | 244 | 6809680 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |