Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162872.1 | NP_001156344.1 | 8 - 9 | 1175 | 67 | |
NM_016857.2 | NP_058553.2 | 9 - 10 | 1268 | 67 | ||
GenBank mRNA | AF014461.1 | - | 9 - 10 | 1203 | 67 | |
AK087601.1 | - | 8 - 9 | 1163 | 67 | ||
AK140450.1 | - | 9 - 10 | 1242 | 67 | ||
AK167769.1 | - | 8 - 9 | 1158 | 67 | ||
AK169928.1 | - | 8 - 9 | 1162 | 67 | ||
AK173094.1 | - | 7 - 8 | 5755 | 67 | ||
BC028927.1 | - | 8 - 9 | 1151 | 4208803 | 67 | |
BC053710.1 | - | 8 - 9 | 1162 | 30056570 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |