Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204275.1 | NP_001191204.1 | 22 - 23 | 3044 | 68 | |
NM_001204276.1 | NP_001191205.1 | 23 - 24 | 3092 | 68 | ||
NM_001204277.1 | NP_001191206.1 | 22 - 23 | 2963 | 68 | ||
NM_001204278.1 | NP_001191207.1 | 21 - 22 | 2933 | 68 | ||
NM_001204279.1 | NP_001191208.1 | 21 - 22 | 2861 | 68 | ||
NM_001204280.1 | NP_001191209.1 | 19 - 20 | 2789 | 68 | ||
NM_001204281.1 | NP_001191210.1 | 20 - 21 | 2496 | 68 | ||
NM_019572.3 | NP_062518.2 | 21 - 22 | 2945 | 68 | ||
GenBank mRNA | AF207749.1 | - | 21 - 22 | 2555 | 68 | |
AK028515.1 | - | 20 - 21 | 2470 | 68 | ||
AK030863.1 | - | 22 - 23 | 2845 | 68 | ||
AK036586.1 | - | 23 - 24 | 2820 | 68 | ||
AK037282.1 | - | 19 - 20 | 2453 | 68 | ||
AK044287.1 | - | 22 - 23 | 2691 | 68 | ||
AK088828.1 | - | 21 - 22 | 2629 | 68 | ||
AK088945.1 | - | 21 - 22 | 2689 | 68 | ||
AK131123.1 | - | 20 - 21 | 3945 | 68 | ||
AK146315.1 | - | 21 - 22 | 2734 | 68 | ||
AK150041.1 | - | 21 - 22 | 2656 | 68 | ||
BC057332.1 | - | 21 - 22 | 2849 | 6826982 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |