Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB048364.1 | - | 3 - 4 | 409 | 64 | |
AK018634.1 | - | 3 - 4 | 236 | 64 | ||
AK086508.1 | - | 3 - 4 | 402 | 64 | ||
BC038274.1 | - | 3 - 4 | 479 | 4457335 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |