Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163729.2 | NP_001157201.2 | 4 - 5 | 738 | 75 | |
NM_027910.3 | NP_082186.2 | 5 - 6 | 891 | 75 | ||
GenBank mRNA | AB053465.1 | - | 5 - 6 | 762 | 75 | |
AJ278127.1 | - | 5 - 6 | 758 | 75 | ||
AK004964.1 | - | 5 - 6 | 868 | 75 | ||
AK141673.1 | - | 4 - 5 | 723 | 75 | ||
AK170623.1 | - | 5 - 6 | 763 | 75 | ||
AK195810.1 | - | 2 - 3 | 142 | 75 | ||
AK201380.1 | - | 2 - 3 | 76 | |||
AK213181.1 | - | 2 - 3 | 142 | 75 | ||
BC018154.1 | - | 5 - 6 | 860 | 3983436 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |