Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028089.3 | NP_082365.1 | 3 - 4 | 513 | 79 | |
GenBank mRNA | AK008580.1 | - | 3 - 4 | 513 | 79 | |
AY206875.1 | - | 3 - 4 | 486 | 79 | ||
BC010824.1 | - | 3 - 4 | 501 | 4219121 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |