Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080470.1 | NP_536718.1 | 2 - 3 | 318 | 82 | |
GenBank mRNA | AF303827.1 | - | 2 - 3 | 318 | 82 | |
BC129800.1 | - | 2 - 3 | 309 | 40048794 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |