Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700019703.mm.1 |
NM_009639.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019703.mm.1 |
NM_009639.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019703.mm.1 |
TC1700002102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019706.mm.1 |
NM_009639.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019706.mm.1 |
NM_009639.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019706.mm.1 |
TC1700002102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019701.mm.1 |
NM_009639.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019701.mm.1 |
NM_009639.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019701.mm.1 |
TC1700002102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019704.mm.1 |
NM_009639.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019704.mm.1 |
NM_009639.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019704.mm.1 |
TC1700002102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019705.mm.1 |
NM_009639.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019705.mm.1 |
NM_009639.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019705.mm.1 |
TC1700002102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019702.mm.1 |
NM_009639.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019702.mm.1 |
NM_009639.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019702.mm.1 |
TC1700002102.mm.1 |
MTA TC |