Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009671.5 | NP_033801.4 | 8 - 9 | 1124 | 58 | |
GenBank mRNA | AB011370.2 | - | 8 - 9 | 1122 | 58 | |
AB098329.1 | - | 8 - 9 | 1122 | 58 | ||
BC139231.1 | - | 8 - 9 | 1135 | 8862253 | 58 | |
BC145583.1 | - | 8 - 9 | 1135 | 9054149 | 58 | |
CV557421.1 | - | 1 - 2 | 151 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |