Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013464.4 | NP_038492.1 | 2 - 3 | 613 | 73 | |
GenBank mRNA | AF405560.1 | - | 2 - 3 | 247 | 73 | |
AF405561.1 | - | 2 - 3 | 247 | 73 | ||
AF405562.1 | - | 2 - 3 | 247 | 73 | ||
AF405563.1 | - | 2 - 3 | 247 | 73 | ||
AF405565.1 | - | 2 - 3 | 247 | 73 | ||
AF405566.1 | - | 2 - 3 | 247 | 73 | ||
AF405567.1 | - | 2 - 3 | 247 | 73 | ||
AF405568.1 | - | 2 - 3 | 247 | 73 | ||
AF405570.1 | - | 2 - 3 | 247 | 73 | ||
AK153678.1 | - | 2 - 3 | 614 | 73 | ||
AK155380.1 | - | 2 - 3 | 665 | 73 | ||
AY662080.1 | - | 2 - 3 | 247 | 73 | ||
AY662082.1 | - | 2 - 3 | 247 | 73 | ||
AY662083.1 | - | 2 - 3 | 247 | 73 | ||
AY662084.1 | - | 2 - 3 | 247 | 73 | ||
D38416.1 | - | 2 - 3 | 262 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |