Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc33a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272035.1 | NP_001258964.1 | 3 - 4 | 1688 | 85 | |
NM_015728.5 | NP_056543.2 | 3 - 4 | 1688 | 85 | ||
GenBank mRNA | AB016795.1 | - | 3 - 4 | 1145 | 85 | |
AK144703.1 | - | 3 - 4 | 1555 | 85 | ||
AW519598.1 | - | 1 - 2 | 167 | 85 | ||
BC054395.1 | - | 3 - 4 | 1688 | 6492236 | 85 | |
BC063104.1 | - | 1621 | 5042025 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |