Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019933.2 | NP_064317.2 | 22 - 23 | 2312 | 117 | |
GenBank mRNA | AF106702.1 | - | 21 - 22 | 2140 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mus musculus Probe spans exons |