Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025383.3 | NP_079659.1 | 4 - 5 | 422 | 63 | |
GenBank mRNA | AK003463.1 | - | 4 - 5 | 389 | 63 | |
AK145874.1 | - | 4 - 5 | 410 | 63 | ||
AK155871.1 | - | 4 - 5 | 410 | 63 | ||
AK156566.1 | - | 4 - 5 | 399 | 63 | ||
AK170987.1 | - | 4 - 5 | 407 | 63 | ||
BC037069.1 | - | 4 - 5 | 421 | 5002765 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |