Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008065.2 | NP_032091.2 | 2 - 3 | 523 | 80 | |
GenBank mRNA | AK139916.1 | - | 2 - 3 | 203 | 80 | |
AK145446.1 | - | 2 - 3 | 203 | 80 | ||
AK145838.1 | - | 2 - 3 | 268 | 80 | ||
AK148087.1 | - | 2 - 3 | 294 | 80 | ||
AK150455.1 | - | 2 - 3 | 203 | 80 | ||
AY282793.1 | - | 2 - 3 | 530 | 80 | ||
AY282794.1 | - | 2 - 3 | 189 | 80 | ||
AY282795.1 | - | 2 - 3 | 405 | 80 | ||
BC052448.1 | - | 2 - 3 | 523 | 6408997 | 80 | |
M74515.1 | - | 2 - 3 | 530 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |