Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008567.1 | NP_032593.1 | 4 - 5 | 712 | 96 | |
GenBank mRNA | AK088493.1 | - | 4 - 5 | 711 | 96 | |
AK145520.1 | - | 4 - 5 | 703 | 96 | ||
AK146272.1 | - | 4 - 5 | 709 | 96 | ||
AK146770.1 | - | 4 - 5 | 704 | 96 | ||
AK146896.1 | - | 4 - 5 | 687 | 96 | ||
AK150726.1 | - | 4 - 5 | 660 | 96 | ||
BC050886.1 | - | 4 - 5 | 720 | 5703922 | 96 | |
BC051658.1 | - | 4 - 5 | 689 | 96 | ||
BC057584.1 | - | 4 - 5 | 690 | 6401827 | 96 | |
D86726.1 | - | 4 - 5 | 712 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |