Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009185.3 | NP_033211.2 | 6 - 7 | 961 | 102 | |
GenBank mRNA | AK049223.1 | - | 6 - 7 | 898 | 102 | |
AK049550.1 | - | 6 - 7 | 898 | 102 | ||
BC049865.1 | - | 6 - 7 | 915 | 102 | ||
BC060706.1 | - | 6 - 7 | 877 | 102 | ||
BC141175.1 | - | 6 - 7 | 834 | 9055787 | 102 | |
BC145493.1 | - | 6 - 7 | 834 | 9054056 | 102 | |
U36778.1 | - | 6 - 7 | 879 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |