Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009791.4 | NP_033921.3 | 20 - 21 | 8176 | 112 | |
GenBank mRNA | AF062378.1 | - | 3 - 4 | 436 | 112 | |
AF533752.1 | - | 20 - 21 | 8154 | 112 | ||
AK048277.1 | - | 3 - 4 | 988 | 112 | ||
AY971958.1 | - | 19 - 20 | 4222 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |