Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009948.2 | NP_034078.2 | 9 - 10 | 1055 | 82 | |
GenBank mRNA | AB010826.1 | - | 8 - 9 | 990 | 82 | |
AF017174.2 | - | 9 - 10 | 1055 | 82 | ||
AK029141.1 | - | 9 - 10 | 1031 | 82 | ||
AK146851.1 | - | 9 - 10 | 1057 | 82 | ||
BC018270.1 | - | 9 - 10 | 1022 | 4920625 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |