Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017861.1 | - | 3 - 4 | 385 | 69 | |
AK146459.1 | - | 4 - 5 | 501 | 69 | ||
AK158704.1 | - | 4 - 5 | 503 | 69 | ||
BC008130.1 | - | 5 - 6 | 506 | 3498984 | 69 | |
BC034861.1 | - | 4 - 5 | 510 | 69 | ||
X71978.1 | - | 4 - 5 | 525 | 69 | ||
Z67963.1 | - | 4 - 5 | 525 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |