Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018761.3 | NP_061231.3 | 1 - 2 | 229 | 97 | |
GenBank mRNA | AF006071.1 | - | 1 - 2 | 216 | 97 | |
AK034393.1 | - | 1 - 2 | 186 | 97 | ||
AK082755.1 | - | 1 - 2 | 172 | 97 | ||
CF896873.1 | - | 1 - 2 | 156 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |