Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019667.2 | NP_062641.1 | 2 - 3 | 174 | 81 | |
GenBank mRNA | AB012611.1 | - | 2 - 3 | 189 | 81 | |
AK004604.2 | - | 2 - 3 | 174 | 81 | ||
AK044230.1 | - | 2 - 3 | 202 | 81 | ||
AK078244.1 | - | 2 - 3 | 206 | 81 | ||
AK159699.1 | - | 2 - 3 | 203 | 81 | ||
AK168729.1 | - | 2 - 3 | 206 | 81 | ||
BC013818.1 | - | 2 - 3 | 210 | 3671426 | 81 | |
DQ141211.1 | - | 3 - 4 | 404 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |